• USA Home
  • Search Results

  • aluminumisopropoxide2042455531711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum isopropoxide

2 Products
Synonym:   AIP, Aluminum isopropylate, Aluminum triisopropoxide
Image of Aluminum isopropoxide
CAS Number:
Linear Formula:
 Al[OCH(CH3)2]3
Molecular Weight:
 204.24
 Beilstein/REAXYS Number:
 3910275
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More