• USA Home
  • Search Results

  • aluminumoxide10196134428111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum oxide

25 Products
Synonym:   Alumina
Image of Aluminum oxide
CAS Number:
Linear Formula:
 Al2O3
Molecular Weight:
 101.96
 EC Number:
 MDL number:
 NACRES:
 SB.52
 PubChem Substance ID:
 eCl@ss:
 38120402
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  Product Line
  Loss
  Density
  Concentration
  Reaction Suitability
  Quality clarifier
  Composite Dimension
  Type
  Manufacturer designation
  Application(s)
  Assay
  Pore size
  pH Value
  ID clarifier
  Surface Dimension
  Special Grade
  Particle Dimension
Show More

Product #
Description
199443 Aluminum oxide, activated, basic, Brockmann I,
199974 Aluminum oxide, activated, neutral, Brockmann I,
544833 Aluminum oxide, nanopowder, <50 nm particle size (TEM),
06300 Aluminum oxide, activated, neutral, Brockmann Activity I,
199966 Aluminum oxide, activated, acidic, Brockmann I,
229423 Aluminum oxide, powder, 99.99% trace metals basis,
718475 Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis,
265497 Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis,
769290 Aluminum oxide, powder, ultra dry,
702129 Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol,
202606 Aluminum oxide, 99.997% trace metals basis,
642991 Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O,
414069 Aluminum oxide, pellets, 3 mm,
234745 Aluminum oxide, Corundum, α-phase, -100 mesh,
A1522 Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I,
267740 Aluminum oxide, pore size 58 Å, ~150 mesh,
551643 Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm,
342718 Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh,
342688 Aluminum oxide, fused, powder, primarily α-phase, -325 mesh,
634875 Aluminum oxide, single crystal substrate, <0001>,
05184 Aluminum oxide, for the determination of hydrocarbons,
342653 Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh,
799300 Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri,
799319 Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri,
67993 Aluminum oxide, for chromatography,