• USA Home
  • Search Results

  • aluminumoxidetlccards1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum oxide/TLC-cards

1 Products
Synonym:   Aluminum TLC Plates: Aluminum with Fluorescence
 EC Number:
 NACRES
 NC.07
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Material
  Feature
Show More