• USA Home
  • Search Results

  • aluminumpotassiumsulfatedodecahydrate47439778424911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aluminum potassium sulfate dodecahydrate

4 Products
Synonym:   Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate
Image of Aluminum potassium sulfate dodecahydrate
CAS Number:
Linear Formula:
 AlK(SO4)2 · 12H2O
Molecular Weight:
 474.39
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Reaction Suitability
  Product Line
  Physical Form
  Special Grade
  pH Value
Show More