• USA Home
  • Search Results

  • amannosidasefromcanavaliaensiformisjackbean12345902542711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Mannosidase from Canavalia ensiformis (Jack bean)

1 Products
Synonym:   α-D-Mannoside mannohydrolase
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.24  ( BRENDA | IUBMB )
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More