• USA Home
  • Search Results

  • amastatinhydrochloridehydrate51101anhydrousbasis10093810111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amastatin hydrochloride hydrate

1 Products
Synonym:   (2S,3R)-3-Amino-2-hydroxy-5-methylhexanoyl-Val-Val-Asp hydrochloride hydrate
Image of Amastatin hydrochloride hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C21H38N4O8 · HCl · xH2O
Molecular Weight:
 511.01 (anhydrous basis)
 Beilstein/REAXYS Number:
 8888370
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More