• USA Home
  • Search Results

  • amentoflavone53846161753411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amentoflavone

3 Products
Synonym:   Didemethyl-ginkgetin, I3′,II8-Biapigenin, Tridemethylsciadopitysin
Image of Amentoflavone
CAS Number:
Empirical Formula (Hill Notation):
 C30H18O10
Molecular Weight:
 538.46
 Beilstein/REAXYS Number:
 380244
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More