• USA Home
  • Search Results

  • amethyldltyrosine1952265848011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Methyl-DL-tyrosine

1 Products
Synonym:   DL-2-Methyl-3-(4-hydroxyphenyl)alanine, AMPT
Image of α-Methyl-<SC>DL</SC>-tyrosine
CAS Number:
Linear Formula:
 4-(HO)C6H4CH2C(CH3)(NH2)CO2H
Molecular Weight:
 195.22
 Beilstein/REAXYS Number:
 2938704
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More