• USA Home
  • Search Results

  • amethylenegbutyrolactone981054765911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Methylene-γ-butyrolactone

1 Products
Synonym:   3-Methylenedihydro-2(3H)-furanone, Tulipane, alpha-Methylene-gamma-butyrolactone
Image of α-Methylene-γ-butyrolactone
CAS Number:
Empirical Formula (Hill Notation):
 C5H6O2
Molecular Weight:
 98.10
 Beilstein/REAXYS Number:
 107939
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More