• USA Home
  • Search Results

  • amethylltyrosine1952267287711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Methyl-L-tyrosine

1 Products
Synonym:   L-2-Methyl-3-(4-hydroxy­phenyl)alanine, L-AMPT
Image of α-Methyl-<SC>L</SC>-tyrosine
CAS Number:
Empirical Formula (Hill Notation):
 C10H13NO3
Molecular Weight:
 195.22
 Beilstein/REAXYS Number:
 2368400
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More