• USA Home
  • Search Results

  • amethylstyrenemonomer118189883911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Methylstyrene monomer

1 Products
Synonym:   2-Phenylpropene, Isopropenylbenzene
Image of α-Methylstyrene monomer
CAS Number:
Linear Formula:
 C6H5C(CH3)=CH2
Molecular Weight:
 118.18
 Beilstein/REAXYS Number:
 969405
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More