• USA Home
  • Search Results

  • aminomethacrylatecopolymer123452493816711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amino methacrylate copolymer

1 Products
Synonym:   2-Methyl-2-propenoic acid butyl ester polymer with 2-(dimethylamino)ethyl 2-methyl-2-propenoate and methyl 2-methyl-2-propenoate
CAS Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More