• USA Home
  • Search Results

  • amiodaronerelatedcompoundh1721086924911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amiodarone Related Compound H

2 Products
Synonym:   2-(Diethylamino)ethyl chloride hydrochloride, 2-Chlorotriethylamine hydrochloride, 2-Chloro-N,N-diethylethylamine hydrochloride
Image of Amiodarone Related Compound H
CAS Number:
Linear Formula:
 (C2H5)2NCH2CH2Cl · HCl
Molecular Weight:
 172.10
 Beilstein/REAXYS Number:
 3591991
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More