• USA Home
  • Search Results

  • amisulpride369487167585911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amisulpride

3 Products
Synonym:   4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide, DAN-2163, Deniban
Image of Amisulpride
CAS Number:
Empirical Formula (Hill Notation):
 C17H27N3O4S
Molecular Weight:
 369.48
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
Show More