• USA Home
  • Search Results

  • amitraz293413308961111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amitraz

3 Products
Synonym:   N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide, NSC 324552
Image of Amitraz
CAS Number:
Empirical Formula (Hill Notation):
 C19H23N3
Molecular Weight:
 293.41
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
Show More