• USA Home
  • Search Results

  • amlodipinebesylate5670511147099611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amlodipine besylate

4 Products
Synonym:   2-[(2-Aminoethoxy)-methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5- pyridinedicarboxylic acid 3-ethyl 5-methyl ester benzene sulfonate, Norvasc
Image of Amlodipine besylate
CAS Number:
Empirical Formula (Hill Notation):
 C20H25ClN2O5 · C6H5SO3H
Molecular Weight:
 567.05
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Anal. Standard Form
  Physical Form
Show More