• USA Home
  • Search Results

  • ammoniasolution1703766441711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonia solution

7 Products
Image of Ammonia solution
CAS Number:
Linear Formula:
 NH3
Molecular Weight:
 17.03
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Application(s)
  Vapor pressure
  Density
  Physical Form
  Assay
Show More