• USA Home
  • Search Results

  • ammonium15n2sulfate134134308658411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium-15N2 sulfate

8 Products
Synonym:   15N Labeled ammonium sulfate, Diazanium-15N2 sulfate
Image of Ammonium-<SUP>15</SUP>N<SUB>2</SUB> sulfate
CAS Number:
Linear Formula:
 (15NH4)2SO4
Molecular Weight:
 134.13
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Application(s)
  Molecular weight
Show More