• USA Home
  • Search Results

  • ammoniumacetate770863161811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium acetate

14 Products
Image of Ammonium acetate
CAS Number:
Linear Formula:
 CH3CO2NH4
Molecular Weight:
 77.08
 Beilstein/REAXYS Number:
 4186741
 EC Number:
 MDL number:
 NACRES:
 NB.21
 PubChem Substance ID:
 eCl@ss
 39021908
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Physical Form
  UV-Absorption Condition
  Product Line
  Vapor pressure
  Application(s)
  Density
  Bulk Density
  Melting Point
  UV-Absorption Values
  Solubility
  pH Value
  Special Grade
Show More

Product #
Description
73594 Ammonium acetate, for mass spectrometry, LiChropur, eluent additive for LC-MS,
A1542 Ammonium acetate, for molecular biology, ≥98%,
431311 Ammonium acetate, ≥99.99% trace metals basis,
238074 Ammonium acetate, ACS reagent, ≥97%,
32301 Ammonium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%,
1.01116 Ammonium acetate, for analysis EMSURE® ACS,Reag. Ph Eur,
5.43834 Ammonium acetate, for HPLC LiChropur,
A7330 Ammonium acetate, BioXtra, ≥98%,
09689 Ammonium acetate, BioUltra, for molecular biology, ≥99.0%,
372331 Ammonium acetate, 99.999% trace metals basis,
A7262 Ammonium acetate, reagent grade, ≥98%,
5.33004 Ammonium acetate, for LC-MS LiChropur,
1.01115 Ammonium acetate, EMPLURA®,
AX1222 Ammonium Acetate, HPLC, Meets ACS Specifications,