• USA Home
  • Search Results

  • ammoniumacetatesolution770863161811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium acetate solution

3 Products
Image of Ammonium acetate solution
CAS Number:
Linear Formula:
 CH3COONH4
Molecular Weight:
 77.08
 Beilstein/REAXYS Number:
 4186741
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
 eCl@ss:
 39021908
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Restriction
  UV-Absorption Values
  Physical Form
  Special Grade
Show More