• USA Home
  • Search Results

  • ammoniumaluminumsulfatedodecahydrate45333778426111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium aluminum sulfate dodecahydrate

1 Products
Synonym:   Aluminum ammonium sulfate dodecahydrate, Ammonium alum
Image of Ammonium aluminum sulfate dodecahydrate
CAS Number:
Linear Formula:
 AlNH4(SO4)2 · 12H2O
Molecular Weight:
 453.33
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  Product Line
  Physical Form
Show More