• USA Home
  • Search Results

  • ammoniumbicarbonate7906106633711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium bicarbonate

3 Products
Synonym:   Ammonium hydrogen carbonate
Image of Ammonium bicarbonate
CAS Number:
Linear Formula:
 NH4HCO3
Molecular Weight:
 79.06
 Beilstein/REAXYS Number:
 4329606
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 38020216
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Special Grade
Show More