• USA Home
  • Search Results

  • ammoniumcarbonate960950687611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium carbonate

3 Products
Synonym:   Hartshorn salt
Image of Ammonium carbonate
CAS Number:
Linear Formula:
 (NH4)2CO3
Molecular Weight:
 96.09
 Beilstein/REAXYS Number:
 4253487
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38020208
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  ID clarifier
  Product Line
  Physical Form
  Special Grade
Show More