• USA Home
  • Search Results

  • ammoniumceriumivnitrate548221677421311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium cerium(IV) nitrate

5 Products
Synonym:   CAN, Ceric ammonium nitrate
Image of Ammonium cerium(IV) nitrate
CAS Number:
Linear Formula:
 Ce(NH4)2(NO3)6
Molecular Weight:
 548.22
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  pH Value
  Purity Grade
  Special Grade
  Physical Form
Show More