• USA Home
  • Search Results

  • ammoniumceriumivsulfatedihydrate632551037847911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium cerium(IV) sulfate dihydrate

4 Products
Synonym:   Ceric ammonium sulfate, Cerium(IV) ammonium sulfate
Image of Ammonium cerium(IV) sulfate dihydrate
CAS Number:
Linear Formula:
 Ce(NH4)4(SO4)4 · 2H2O
Molecular Weight:
 632.55
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Reaction Suitability
  Physical Form
Show More