• USA Home
  • Search Results

  • ammoniumchloride53491212502911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium chloride

16 Products
Synonym:   Ammonium chloride, Sal ammoniac
Image of Ammonium chloride
CAS Number:
Empirical Formula (Hill Notation):
 H4ClN
Molecular Weight:
 53.49
 EC Index Number
 235-186-4
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Suitability
  UV-Absorption Values
  Application(s)
  Potency
  Density
  Bulk Density
  Autoignition temp
  Agency/Method
  Vapor pressure
  Solubility
  Purity Grade
  Melting Point
  Physical Form
  pH Value
  Assay
Show More

Product #
Description
A9434 Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%,
254134 Ammonium chloride, 99.998% trace metals basis,
213330 Ammonium chloride, ACS reagent, ≥99.5%,
326372 Ammonium chloride, 99.99% trace metals basis,
09718 Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT),
31107 Ammonium chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%,
A4514 Ammonium chloride, ReagentPlus®, ≥99.5%,
11209 Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance),
09724 Ammonium chloride, tested according to Ph Eur,
1.01145 Ammonium chloride, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
1.01142 Ammonium chloride, EMPROVE® ESSENTIAL Ph Eur,BP,USP,
1.01143 Ammonium chloride, 99.995 Suprapur®,
PHR1327 Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material,
901230 Ammonium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%,
1029953 Ammonium chloride, United States Pharmacopeia (USP) Reference Standard,
AX1270 Ammonium Chloride, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,