• USA Home
  • Search Results

  • ammoniumglycyrrhizate839965395604011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium glycyrrhizate

3 Products
Synonym:   Glycyrrhizic acid ammonium salt
Image of Ammonium glycyrrhizate
CAS Number:
Empirical Formula (Hill Notation):
 C42H62O16 · NH3
Molecular Weight:
 839.96
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More