• USA Home
  • Search Results

  • ammoniumhexachloroosmateiv439021212508511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium hexachloroosmate(IV)

2 Products
Synonym:   Ammonium osmium chloride, Osmium(IV)-ammonium chloride
Image of Ammonium hexachloroosmate(IV)
CAS Number:
Linear Formula:
 (NH4)2OsCl6
Molecular Weight:
 439.02
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More