• USA Home
  • Search Results

  • ammoniumionsolutionforise53491212502911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium ion solution for ISE

1 Products
Synonym:   Ammonium chloride solution
Image of Ammonium ion solution for ISE
CAS Number:
Linear Formula:
 NH4Cl
Molecular Weight:
 53.49
 MDL number:
 NACRES:
 NB.61
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Concentration
Show More