• USA Home
  • Search Results

  • ammoniumironiisulfatehexahydrate39214778385911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium iron(II) sulfate hexahydrate

6 Products
Synonym:   Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt
Image of Ammonium iron(II) sulfate hexahydrate
CAS Number:
Linear Formula:
 (NH4)2Fe(SO4)2 · 6H2O
Molecular Weight:
 392.14
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Solubility
  Reaction Suitability
  Special Grade
  Product Line
  pH Value
  Physical Form
Show More