• USA Home
  • Search Results

  • ammoniumlaurylsulfatesolution28343223554311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium lauryl sulfate solution

1 Products
Synonym:   Ammonium dodecyl sulfate, Dodecyl sulfate ammonium salt
Image of Ammonium lauryl sulfate solution
CAS Number:
Linear Formula:
 CH3(CH2)11OSO3NH4
Molecular Weight:
 283.43
 Beilstein/REAXYS Number:
 5188379
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Density
Show More