• USA Home
  • Search Results

  • ammoniummetatungstatehydrate295630anhydrousbasis1233311811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium metatungstate hydrate

3 Products
Image of Ammonium metatungstate hydrate
CAS Number:
Linear Formula:
 (NH4)6H2W12O40 · xH2O
Molecular Weight:
 2956.30 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More