• USA Home
  • Search Results

  • ammoniummolybdate1235861205485211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium molybdate

1 Products
Synonym:   Ammonium heptamolybdate tetrahydrate, Ammonium molybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Image of Ammonium molybdate
CAS Number:
Linear Formula:
 (NH4)6Mo7O24 · 4H2O
Molecular Weight:
 1235.86
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More