• USA Home
  • Search Results

  • ammoniummolybdatetetrahydrate1235861205485211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium molybdate tetrahydrate

4 Products
Synonym:   Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Image of Ammonium molybdate tetrahydrate
CAS Number:
Linear Formula:
 (NH4)6Mo7O24 · 4H2O
Molecular Weight:
 1235.86
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Product Line
  Special Grade
Show More