• USA Home
  • Search Results

  • ammoniumnickeliisulfatehexahydrate39499778520811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium nickel(II) sulfate hexahydrate

3 Products
Synonym:   Nickel ammonium sulfate
Image of Ammonium nickel(II) sulfate hexahydrate
CAS Number:
Linear Formula:
 (NH4)2Ni(SO4)2 · 6H2O
Molecular Weight:
 394.99
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Reaction Suitability
Show More