• USA Home
  • Search Results

  • ammoniumnitrate15n81043143246911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium nitrate-15N

4 Products
Synonym:   15N ammonium salt, 15N labeled ammonium nitrate
Image of Ammonium nitrate-<SUP>15</SUP>N
CAS Number:
Linear Formula:
 NH415NO3
Molecular Weight:
 81.04
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Molecular weight
Show More