• USA Home
  • Search Results

  • ammoniumnitrate8004648452211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium nitrate

8 Products
Image of Ammonium nitrate
CAS Number:
Linear Formula:
 NH4NO3
Molecular Weight:
 80.04
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 38060309
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Potency
  Melting Point
  Density
  Bulk Density
  Solubility
  Purity Grade
  pH Value
  Special Grade
Show More