• USA Home
  • Search Results

  • ammoniumpersulfate22820772754011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium persulfate

6 Products
Synonym:   AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER
Image of Ammonium persulfate
CAS Number:
Linear Formula:
 (NH4)2S2O8
Molecular Weight:
 228.20
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 38030421
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Purity Grade
  Product Line
  Restriction
  Solubility
  Assay
Show More