• USA Home
  • Search Results

  • ammoniumphosphatemonobasic11503772276111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium phosphate monobasic

8 Products
Synonym:   mono-Ammonium phosphate, prim-Ammonium phosphate, APM, Ammonium dihydrogenphosphate
Image of Ammonium phosphate monobasic
CAS Number:
Linear Formula:
 NH4H2PO4
Molecular Weight:
 115.03
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Suitability
  UV-Absorption Values
  Application(s)
  pH Value
  Solubility
  Assay
Show More