• USA Home
  • Search Results

  • ammoniumpyrrolidinedithiocarbamate16429510896311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium pyrrolidinedithiocarbamate

2 Products
Synonym:   1-Pyrrolidinecarbodithioic acid ammonium salt, APDC, Ammonium pyrrolidinecarbodithioate, PDC, PDTC
Image of Ammonium pyrrolidinedithiocarbamate
CAS Number:
Linear Formula:
 C5H9NS2 · NH3
Molecular Weight:
 164.29
 Beilstein/REAXYS Number:
 3730472
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More