• USA Home
  • Search Results

  • ammoniumsulfamate11412777306011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium sulfamate

3 Products
Image of Ammonium sulfamate
CAS Number:
Linear Formula:
 NH4SO3NH2
Molecular Weight:
 114.12
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
 eCl@ss:
 38060607
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Solubility
  Product Line
  Special Grade
  pH Value
  Assay
Show More