• USA Home
  • Search Results

  • ammoniumsulfitesolution116141019604011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium sulfite solution

1 Products
Image of Ammonium sulfite solution
CAS Number:
Linear Formula:
 (NH4)2SO3
Molecular Weight:
 116.14
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More