• USA Home
  • Search Results

  • ammoniumthiocyanate7612176295411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ammonium thiocyanate

5 Products
Synonym:   Ammonium rhodanide
Image of Ammonium thiocyanate
CAS Number:
Linear Formula:
 NH4SCN
Molecular Weight:
 76.12
 Beilstein/REAXYS Number:
 3595135
 EC Number:
 MDL number:
 NACRES:
 NB.24
 PubChem Substance ID:
 eCl@ss:
 38060604
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Solubility
  Agency/Method
  Product Line
  Vapor pressure
  Physical Form
  Special Grade
  pH Value
Show More