• USA Home
  • Search Results

  • amoxapine313781402844511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amoxapine

4 Products
Synonym:   2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine
Image of Amoxapine
CAS Number:
Empirical Formula (Hill Notation):
 C17H16ClN3O
Molecular Weight:
 313.78
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Package quantity
  Manufacturer designation
Show More