• USA Home
  • Search Results

  • amphetamined5deuteriumlabelonsidechainsolution1402413676527011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Amphetamine-d5 (deuterium label on side chain) solution

2 Products
Image of (±)-Amphetamine-d<SUB>5</SUB> (deuterium label on side chain) solution
CAS Number:
Empirical Formula (Hill Notation):
 C9D5H8N
Molecular Weight:
 140.24
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More