• USA Home
  • Search Results

  • amphetamined6solution141247375826611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Amphetamine-d6 solution

2 Products
Image of (±)-Amphetamine-d<SUB>6</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C9D6H7N
Molecular Weight:
 141.24
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More