• USA Home
  • Search Results

  • amphotericinb92408139789311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amphotericin B

3 Products
CAS Number:
Empirical Formula (Hill Notation):
 C47H73NO17
Molecular Weight:
 924.08
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
Show More