• USA Home
  • Search Results

  • amphotericinbfromstreptomycessp92408139789311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amphotericin B from Streptomyces sp.

2 Products
Synonym:   Fungizone
Image of Amphotericin B from <I>Streptomyces</I> sp.
CAS Number:
Empirical Formula (Hill Notation):
 C47H73NO17
Molecular Weight:
 924.08
 Beilstein/REAXYS Number:
 78342
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Special Grade
  Titertest
  Color
  Biological Source
  Mode of Action
  Assay
  Solubility
Show More