• USA Home
  • Search Results

  • amphotericinbsolution92408139789311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amphotericin B solution

1 Products
Image of Amphotericin B solution
CAS Number:
Empirical Formula (Hill Notation):
 C47H73NO17
Molecular Weight:
 924.08
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More