• USA Home
  • Search Results

  • ampicillin40345717748211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ampicillin

1 Products
Synonym:   D-(−)-α-Aminobenzylpenicillin, Ampicillin trihydrate
Image of Ampicillin
CAS Number:
Empirical Formula (Hill Notation):
 C16H19N3O4S · 3H2O
Molecular Weight:
 403.45
 Beilstein Registry Number:
 5399534
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More